Clobetasone butyrate ≥98%

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 21-Chloro-9-fluoro-17-hydroxy-16-methylpregna-1,4-diene-3,11,20-trione 17-butyrate

M-Clarity™ Qualitätsstufe = MQ200

C5548-100MG
C5548-100MGEA 141 EUR
C5548-100MG
Clobetasone butyrate ≥98%
Clobetasone butyrate

Clobetasone butyrate is a corticosteroid. It can be used as a diagnostic assay manufacturing, hematology, histology. Clobetasone butyrate has been used as an internal standard for the quantitative assay of clobetasol propionate. Clobetasone butyrate has been used to analyse its bio-concentration factor (BCF) in cyprinus carpio. Clobetasone butyrate is a potent topical corticosteroid and is primarily used to treat atopic dermatitis (AD). Low dosage of this drug is used to treat dry eye in sjogrens syndrome.

Formel: C₂₆H₃₂ClFO₅
Molecular Weight: 478,98 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00133196
CAS-Nummer: 25122-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR