Chlorophyll A, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

C6144-1MG C6144-5MG
C6144-1MGEA 260 EUR
C6144-1MG C6144-5MG
Chlorophyll A, Sigma-Aldrich®
Chlorophyll A

Chlorophyll a has been used as a standard for the calibration of fluorometer.

Formel: C₅₅H₇₂MgN₄O₅
Molecular Weight: 893.49 g/mol
Schmelzpunkt: 150…153 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00079050
CAS-Nummer: 479-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd945840481c47eeb5edc67c3406ddc48f34bb0c&vid=628e59622c8ba54182dffa4d51a091297007b43f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR