N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline ≥95% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: CDCQ

C6206-25MG
C6206-25MGEA 1270 EUR
C6206-25MG
N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline ≥95% (durch HPLC), Sigma-Aldrich®
N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline

N-Carboxymethyl-6-(2,2-dicyanovinyl)-1,2,3,4-tetrahydroquinoline. ≥98% (HPLC) (CDCQ)

Formel: C₁₅H₁₃N₃O₂
Molecular Weight: 267,28 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00274409
CAS-Nummer: 47072-52-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b3946e309033154cb20c90ebab78005ca947fad&vid=70fc69dc8418d5789b1bff2cd11f49ec7b37d182&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR