9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N-hydroxysuccinimide ester chloride, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: NIR-1

C6331-1MG
C6331-1MGEA 1270 EUR
C6331-1MG
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N-hydroxysuccinimide ester chloride, Sigma-Aldrich®
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N-hydroxysuccinimide ester chloride

Fluorescent label for amino acids allows for detection of alanine, arginine, glycine, glutamic and aspartic acid in the 0.8-4.5 × 10-18 M range.

Formel: C₃H₃₅ClN₄O₅
Molecular Weight: 567,08 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01310495
CAS-Nummer: 158358-57-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a29617fdebf6f73eb50fdc269175ea2bb24d8c7&vid=48b2b0e573d6822887b3d4095ae52c0a30277d83&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR