9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N⁴-(maleimidoethyl)piperazide chloride, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

C6456-1MG
C6456-1MGEA 1270 EUR
C6456-1MG
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N⁴-(maleimidoethyl)piperazide chloride, Sigma-Aldrich®
9-Cyano-N,N,N′-triethylpyronine-N′-caproic acid N⁴-(maleimidoethyl)piperazide chloride

Fluorescent protein label.

Formel: C₃₆H₄₅ClN₆O₄
Lagertemperatur: Gefrierschrank
MDL: MFCD01310494
CAS-Nummer: 163032-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1901ee75effd0fe9c0afe3e8b5d41bc2a4d5bad9&vid=0db9cd9e75f62d8147cb88833475ac0352b60497&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR