Organic reference standard, Casein from bovine milk, for use in the Total Dietary Fiber Control Kit, TDF-C10, Supelco®
Lieferant: Merck
Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.
Verpackung: 5 g in poly bottle.