5(6)-Carboxyfluorescein diacetate Gemisch aus Isomeren ≥90% (durch HPLC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
CFDA, 5(6)-Carboxy-di-O-acetylfluorescein, 5(6)-CFDA
C8166-100MG
C8166-25MG
C8166-100MGEA
356
EUR
C8166-100MG
C8166-25MG
5(6)-Carboxyfluorescein diacetate Gemisch aus Isomeren ≥90% (durch HPLC), Sigma-Aldrich®
5(6)-Carboxyfluorescein diacetate
5(6)-Carboxyfluorescein diacetate Mixed isomers. ≥90% (HPLC)
Formel:
C₂₅H₁₆O₉ Molecular Weight: 460.39 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00467494 CAS-Nummer: 124387-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...