Crocein scarlet 7B, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
C.I. 27165, Acid Red 71
C8822-1G
C8822-25G
C8822-5G
C8822-1GEA
41.3
EUR
C8822-1G
C8822-25G
C8822-5G
Crocein scarlet 7B, Sigma-Aldrich®
Crocein scarlet 7B
Crocein Scarlet 7B, (Acid Red 71)
Formel:
C₂₄H₁₈N₄Na₂O₇S₂ Molecular Weight: 584.53 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056588 CAS-Nummer: 6226-76-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...