trans-4-Hydroxyzimtsäure ≥98% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

C9008-25G C9008-10G C9008-1G C9008-5G
C9008-25GEA 112 EUR
C9008-25G C9008-10G C9008-1G C9008-5G
trans-4-Hydroxyzimtsäure ≥98% (durch HPLC), Sigma-Aldrich®
trans-4-Hydroxyzimtsäure

p-Coumaric acid has been used as a component of chemiluminescent substrate for protein detection in western blotting.


  • In western blot, p-coumaric is also used as an enhancer in chemiluminescent substrate
  • Hydroxycinnamic acid found in many fruits and vegetables
  • Predominantly trans isomer

Formel: C₉H₈O₃
Schmelzpunkt: 214 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00004399
CAS-Nummer: 501-98-4
EINECS: 231-000-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4645cd3c9d127cab2ae6da450e82710d89ca9d8f&vid=7c075fda0e75e2d1e4bc45e5e4ab7dd0ea62103e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow and Faint Beige to Beige and Faint Brown to Brown
Appearance (Form) Powder
Purity (Hplc Area %) ≥98.0 %
Solubility (Colour) Colorless to Yellow
Solubility (Turbidity) Clear
Solubility (Method) 50 mg/ml, Ethanol
Proton Nmr Spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR