Bis-(cyclohexanon)-oxaldihydrazon, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone
C9012-25G
C9012-25GEA
74
EUR
C9012-25G
Bis-(cyclohexanon)-oxaldihydrazon, Sigma-Aldrich®
Bis(cyclohexanone)oxalyldihydrazone
Bis(cyclohexanone)oxaldihydrazone (Cuprizone) has been used in mice to study the effect of NG2 (neural/glial antigen 2) proteoglycan on toxic-induced demyelination. It has been used to induce demyelination in mice. It has been used to study cuprizone-induced demyelination and remyelination in the corpus callosum of mice.
Formel:
C₁₄H₂₂N₄O₂ Molecular Weight: 278,36 g/mol Schmelzpunkt: 210…214 °C (Lit.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001659 CAS-Nummer: 370-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...