Bunsenbrenner, Labogaz® 470
Lieferant: CAMPING GAZ
2000020657
CAMP2000020657EA 110 EUR
CAMP2000020657
Bunsenbrenner, Labogaz® 470
Heizgeräte Gasbrenner
Compact bunsen burner for operation with the Campingaz cartridge CV 470 Plus.
  • Device can be removed from the cartridge even if the cartridge is not yet completely empty
  • Ideal for heating solutions in glass flasks and test tubes or bending parts such as glass tubes
For a wide range of applications in crafts (optics, goldsmith work), industrial laboratories (physics, chemistry, pharmaceuticals) or schools (practical work, experiments).