Blow Lamp, X 1650
Lieferant: CAMPING GAZ
2000026867
CAMP2000026867EA 22.4 EUR
CAMP2000026867
Blow Lamp, X 1650
Heizgeräte Gasbrenner
Extra fine flame burner. Ideal for any precise work such as soldering small components, jewelry or models
  • Easy screw attachment
  • Gas consumption: 30 g/h