Gas Burner, Bleuet Micro Plus
Lieferant: CAMPING GAZ
2184543
CAMP2184543EA 32.7 EUR
CAMP2184543
Gas Burner, Bleuet Micro Plus
Heizgeräte Gasbrenner
  • Maximum power: 1250 W
  • With Easy-clic Plus system
  • Four foldable side arms
  • Gas cartridge must be ordered separately
Consumption: 90 g/hour
Weight: 180 g
Bestellinformation: Supplied without gas cartridges, cartridges CV300 plus or CV470 plus must be ordered separately.