for removing of antibodies from Western blotting membranes for reprobing on the same membrane, non-malodorant and non-hazardous because free from DTT and mercaptoethanol, no need for a fume hood, ready-to-use 1 * 50 mL
Lieferant: CANDOR BIOSCIENCE
for removing of antibodies from Western blotting membranes for reprobing on the same membrane, non-malodorant and non-hazardous because free from DTT and mercaptoethanol, no need for a fume hood, ready-to-use 1 * 50 mL