Viscosity standards, general purpose
Lieferant: CANNON INSTRUMENT
9727-C45
9727-C60
9727-C65
9727-C25.016
CANN9727-C45EA
0
EUR
CANN9727-C45
CANN9727-C60
CANN9727-C65
CANN9727-C25.016
Viscosity standards, general purpose
Standards
Viskosimetrie-Standards
These certified standards are useful for calibration and verification procedures.
- It is suitable for all types of viscometers
- Viscosity data quoted at 20 °C
- It also helps ensures the reliability of measurements
Standards: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.