Viscosity standards, general purpose
Lieferant: CANNON INSTRUMENT
9727-C45 9727-C60 9727-C65 9727-C25.016
CANN9727-C45EA 0 EUR
CANN9727-C45 CANN9727-C60 CANN9727-C65 CANN9727-C25.016
Viscosity standards, general purpose
Standards Viskosimetrie-Standards
These certified standards are useful for calibration and verification procedures.
  • It is suitable for all types of viscometers
  • Viscosity data quoted at 20 °C
  • It also helps ensures the reliability of measurements
Standards: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.