Silicone Viscosoty Standards for Rotational Viscometers
Lieferant: CANNON INSTRUMENT
9727-Z30 9727-Z34
CANN9727-Z30EA 568 EUR
CANN9727-Z30 CANN9727-Z34
Silicone Viscosoty Standards for Rotational Viscometers
Standards Viskosimetrie-Standards
CANNON silicone viscosity standards are intended for use with rotational viscometers only.
Silicone viscosity standards are not recommended for glass capillary viscometers (including vacuum viscometers), metal cup-type viscometers (such as ISO, Zahn, Ford and Shell cups), or any other viscometers providing kinematic viscosity—CANNON hydrocardon-based oil standards should be used for these types of viscometers.
Viscosity standards are traceable to a NIST calibration and prepared in Cannon's ISO 9001-registered, A2LA-accredited laboratory.
Standards: Supplied with certificate.
Bitte beachten Viscosities quoted in the table below are nominal. Lot specific viscosity data in mPa·s (cP) at 20 °C, 23 °C, 24 °C, 25 °C, 26 °C, 27 °C and 40 °C is provided with each standard.