Protein carbonyl colorimetric assay kit
Lieferant: Cayman Chemical
10005020-96
CAYM10005020-96EA 424 EUR
CAYM10005020-96
Protein carbonyl colorimetric assay kit
Tests Proteintests
The Protein Carbonyl Colorimetric Assay Kit is for the measurement of oxidised proteins. Protein samples are derivatised by making use of the reaction between 2,4-dinitrophenylhydrazine and protein carbonyls.
  • Measure protein carbonyl content in plasma, serum, urine, tissue homogenates and cell lysates
  • Assay 24 samples in duplicate
  • Plate-based colorimetric measurement (360 to 385 nm)