Human recombinant Renin
Lieferant: Cayman Chemical
10006217-25
CAYM10006217-25EA
509
EUR
CAYM10006217-25
Human recombinant Renin
Proteine und Peptide
Renin is an aspartyl protease of approximately 40 kDa produced in the kidney by juxtaglomerular cells of the macula densa.
- Renin converts angiotensinogen into angiotensin I
- Mature and active
- 340 amino acid protease
Native human renin is a glycoprotein derived from preprorenin by sequential cleavage of 20 and 46 amino acid residues from the N-terminus of the 406 amino acid precursor.