PPARα transcription factor assay kit
Lieferant: Cayman Chemical
A 96-well assay for measurement of PPARα DNA binding activity.
- Sensitive, non radioactive method of detecting PPARα from whole cell lysates
- 96-well plate format replaces EMSAs
- Capture the transcription factor using a specific dsDNA sequence bound to the plate
- Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
PPARs are ligand-activated transcription factors belonging to the large superfamily of nuclear receptors. PPARα primarily activates genes encoding proteins involved in fatty acid oxidation, while PPARγ primarily activates genes directly involved in lipogenic pathway and insulin signaling.
Members of the PPAR family are important direct targets of many antidiabetic and hypolipidemic drugs. PPAR Transcription Factor Assays are a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
Lieferumfang: Kit contains: Assay buffer, Reagent A, positive control, binding buffer, antibody, concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover, developing and stop solution.