Indol-4-carbonitril

Lieferant: Cayman Chemical
Achtung

Synonyme: 4-Indolcarbonitril, 4-Cyanindol, 1H-Indol-4-carbonitril, 4-Cyan-1H-indol

10007084-1 10007084-10 10007084-5 10007084-500
CAYM10007084-1EA 99 EUR
CAYM10007084-1 CAYM10007084-10 CAYM10007084-5 CAYM10007084-500
Indol-4-carbonitril
Indol-4-carbonitril
Formel: C₉H₆N₂
Molecular Weight: 142.16 g/mol
MDL: MFCD00152045
CAS-Nummer: 16136-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR