2-Aminopyrazin
Lieferant: Cayman Chemical
Synonyme:
Aminopyrazin, Pyrazin-2-amin
10007093-5
10007093-10
10007093-25
10007093-50
CAYM10007093-5EA
33.8
EUR
CAYM10007093-5
CAYM10007093-10
CAYM10007093-25
CAYM10007093-50
2-Aminopyrazin
2-Aminopyrazine
Formel:
C₄H₅N₃ Siedepunkt: 203 °C (1013 hPa) Schmelzpunkt: 117…120 °C |
MDL:
MFCD00006137 CAS-Nummer: 5049-61-6 EINECS: 225-748-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...