2-Pyrimidincarbonitril
Lieferant: Cayman Chemical
Synonyme:
2-Cyanopyrimidine
10007121-1
10007121-10
10007121-5
10007121-500
CAYM10007121-1EA
26
EUR
CAYM10007121-1
CAYM10007121-10
CAYM10007121-5
CAYM10007121-500
2-Pyrimidincarbonitril
2-Pyrimidincarbonitril
Formel:
C₅H₃N₃ Molecular Weight: 105.1 g/mol |
MDL:
MFCD00160513 CAS-Nummer: 14080-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...