2-Pyrimidincarbonitril

Lieferant: Cayman Chemical

Synonyme: 2-Cyanopyrimidine

10007121-1 10007121-10 10007121-5 10007121-500
CAYM10007121-1EA 26 EUR
CAYM10007121-1 CAYM10007121-10 CAYM10007121-5 CAYM10007121-500
2-Pyrimidincarbonitril
2-Pyrimidincarbonitril
Formel: C₅H₃N₃
Molecular Weight: 105.1 g/mol
MDL: MFCD00160513
CAS-Nummer: 14080-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR