tert-Butyl-4-(brommethyl)benzoat

Lieferant: Cayman Chemical

Synonyme: 4-Bromo-methyl-benzoic acid mono tert-butyl ester, tert-Butyl 4-(bromomethyl)benzoate

10007129-1
CAYM10007129-1EA 95 EUR
CAYM10007129-1
tert-Butyl-4-(brommethyl)benzoat
tert-Butyl-4-(brommethyl)benzoat
Formel: C₁₂H₁₅BrO₂
MDL: MFCD04973450
CAS-Nummer: 108052-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR