Ethyllinoleat

Lieferant: Cayman Chemical

10008198-100 10008198-50 10008198-500 10008198-1
CAYM10008198-100EA 27 EUR
CAYM10008198-100 CAYM10008198-50 CAYM10008198-500 CAYM10008198-1
Ethyllinoleat
Ethyllinoleat
Formel: C₂₀H₃₆O₂
Lagertemperatur: Gefrierschrank
MDL: MFCD00009535
CAS-Nummer: 544-35-4
EINECS: 208-868-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR