di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP), Tabletten
Lieferant: Cayman Chemical
10009147-1
CAYM10009147-1EA
97
EUR
CAYM10009147-1
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP), Tabletten
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
Acid Phosphatase Substrate
Formel:
C₆H₄NNa₂O₆P·6H₂O Schmelzpunkt: 300 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00066288 CAS-Nummer: 333338-18-4 EINECS: 224-246-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...