Anti-PSD95 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
10009506-1
CAYM10009506-1EA 645 EUR
CAYM10009506-1
Anti-PSD95 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PSD95
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR