3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid

Lieferant: Cayman Chemical
Achtung

Synonyme: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

10009591-1 10009591-5
CAYM10009591-1EA 57 EUR
CAYM10009591-1 CAYM10009591-5
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formel: C₁₈H₁₆N₅BrS
Lagertemperatur: Kühlschrank
MDL: MFCD00011964
CAS-Nummer: 298-93-1
EINECS: 206-069-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR