Anti-EP4 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
101770-1
CAYM101770-1EA 418 EUR
CAYM101770-1
Anti-EP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EP4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR