Prostaglandin Screening Library I
Lieferant: Cayman Chemical
10501-50
CAYM10501-50EA 1999 EUR
CAYM10501-50
Prostaglandin Screening Library I
Tests Zelltests
For screening of PGF analogs.
  • Contains >70 compounds
  • Includes the ''F-series'' prostaglandins and isoprostanes
The Prostaglandin Screening Library I contains more than 70 ''F-series'' of prostaglandins and isoprostanes in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.