Fatty Acid Screening Library
Lieferant: Cayman Chemical
10504-50
CAYM10504-50EA 1958 EUR
CAYM10504-50
Fatty Acid Screening Library
Tests Zelltests
For screening of variety of fatty acids.
  • Contains >75 compounds
  • Includes arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids
The Fatty Acid Screening Library contains more than 75 compounds of fatty acids with diverse biological activities in a 96-well Matrix tube rack format as 2 mM stocks in DMSO. The library may also include some of the most commonly-ordered fatty acids, including arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids. As well as some unusual fatty acids, including 9-thiastearic acid, 2-hydroxymyristic acid, AUDA, and CUDA may be included. The composition of this screening library will always vary somewhat depending upon our inventory. Stability data is not available for compounds as supplied in the screening library.