Dexamethason

Lieferant: Cayman Chemical

Synonyme: 9-Fluor-11β,17α,21-trihydroxy-16α-methyl-pregna-1,4-dien-3,20-dion, Dexamethasonum

11015-500 11015-1 11015-5
CAYM11015-500EA 76 EUR
CAYM11015-500 CAYM11015-1 CAYM11015-5
Dexamethason
Dexamethason
Formel: C₂₂H₂₉FO₅
Schmelzpunkt: 268…271 °C
MDL: MFCD00064136
CAS-Nummer: 50-02-2
EINECS: 200-003-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR