3-Aminobenzamidoxim

Lieferant: Cayman Chemical

11074-500 11074-1 11074-5
CAYM11074-500EA 43 EUR
CAYM11074-500 CAYM11074-1 CAYM11074-5
3-Aminobenzamidoxim
3-Aminobenzamidoxim
Formel: C₇H₉N₃O
MDL: MFCD08061150
CAS-Nummer: 100524-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR