7-Aminoactinomycin (7-AAD)

Lieferant: Cayman Chemical

11397-1 11397-5
CAYM11397-1EA 146 EUR
CAYM11397-1 CAYM11397-5
7-Aminoactinomycin (7-AAD)
7-Aminoactinomycin D (7-AAD)
Formel: C₆₂H₈₇N₁₃O₁₆
MDL: MFCD00077295
CAS-Nummer: 7240-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR