Kämpferol
Lieferant: Cayman Chemical
Synonyme:
3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
11852-25
11852-50
11852-100
CAYM11852-25EA
59
EUR
CAYM11852-25
CAYM11852-50
CAYM11852-100
Kämpferol
Kämpferol
Formel:
C₁₅H₁₀O₆ Molecular Weight: 286.24 g/mol |
MDL:
MFCD00016938 CAS-Nummer: 520-18-3 EINECS: 208-287-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...