2-Methylbenzamidoxim

Lieferant: Cayman Chemical

13002-500 13002-1 13002-5
CAYM13002-500EA 42 EUR
CAYM13002-500 CAYM13002-1 CAYM13002-5
2-Methylbenzamidoxim
2-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
CAS-Nummer: 40312-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR