4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure

Lieferant: Cayman Chemical

13047-100 13047-250 13047-500 13047-1
CAYM13047-100EA 22 EUR
CAYM13047-100 CAYM13047-250 CAYM13047-500 CAYM13047-1
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Formel: C₁₀H₈N₂O₂S
Molecular Weight: 220.25 g/mol
MDL: MFCD01316364
CAS-Nummer: 34418-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR