4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Lieferant: Cayman Chemical
13047-100
13047-250
13047-500
13047-1
CAYM13047-100EA
22
EUR
CAYM13047-100
CAYM13047-250
CAYM13047-500
CAYM13047-1
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
4-Methyl-2-(2-pyridinyl)-5-thiazolcarbonsäure
Formel:
C₁₀H₈N₂O₂S Molecular Weight: 220.25 g/mol |
MDL:
MFCD01316364 CAS-Nummer: 34418-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...