Ethyl-2-amino-4-methylthiazol-5-carboxylat
Lieferant: Cayman Chemical
Synonyme:
Ethyl 2-amino-4-methylthiazole-5-carboxylate
13059-250
13059-500
13059-1
13059-5
CAYM13059-250EA
19
EUR
CAYM13059-250
CAYM13059-500
CAYM13059-1
CAYM13059-5
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel:
C₇H₁₀N₂O₂S |
MDL:
MFCD00123414 CAS-Nummer: 7210-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...