Ethyl-2-amino-4-methylthiazol-5-carboxylat

Lieferant: Cayman Chemical

Synonyme: Ethyl 2-amino-4-methylthiazole-5-carboxylate

13059-250 13059-500 13059-1 13059-5
CAYM13059-250EA 19 EUR
CAYM13059-250 CAYM13059-500 CAYM13059-1 CAYM13059-5
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Ethyl-2-amino-4-methylthiazol-5-carboxylat
Formel: C₇H₁₀N₂O₂S
MDL: MFCD00123414
CAS-Nummer: 7210-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR