Streptozocin
Lieferant: Cayman Chemical
|
Gefahr
|
13104-50
13104-1
CAYM13104-50EA
32
EUR
CAYM13104-50
CAYM13104-1
Streptozocin
Streptozocin
Formel:
C₈H₁₅N₃O₇ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006607 CAS-Nummer: 18883-66-4 EINECS: 242-646-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...