Human thrombin (from blood)
Lieferant: Cayman Chemical
13188-1
CAYM13188-1EA 633 EUR
CAYM13188-1
Human thrombin (from blood)
Enzyme
Thrombin is the last enzyme in the clotting cascade functioning to cleave fibrinogen to fibrin and potentiates the procoagulation process by activating factors V, VIII, XI, and XIII.
Thrombin aids the anticoagulant process by activating protein C. Activated protein C inhibits activated factors V and VIII ultimately down-regulating further thrombin production. Thrombin is often used as a potent platelet activator in vitro. Platelet responses to thrombin are mediated through protease-activated receptors (PARs). Thrombin-mediated signaling regulates a diverse range of metabolic processes including activation of phospholipases in platelets, endothelial cells, and cardiac myocytes.
Formulated as 5 mM sodium citrate, pH 6,5, containing 0,01% PEG and 20 mM sodium chloride.