3-Methyl-1-phenyl-2-pyrazolin-5-on
Lieferant: Cayman Chemical
Synonyme:
MCI-186, 1-Phenyl-3-methyl-5-pyrazolone, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one, Edaravone
13320-500
CAYM13320-500EA
59
EUR
CAYM13320-500
3-Methyl-1-phenyl-2-pyrazolin-5-on
3-Methyl-1-phenyl-2-pyrazolin-5-on
Formel:
C₁₀H₁₀N₂O Molecular Weight: 174,2 g/mol |
MDL:
MFCD00003138 CAS-Nummer: 89-25-8 EINECS: 201-891-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...