3-Isobutyl-1-methylxanthin

Lieferant: Cayman Chemical
Achtung

Synonyme: 3-Isobutyl-1-methyl-2,6(1H,3H)-purindion, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-3,7-dihydro-2H-purin-2,6-dion, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purin-2,6-dion, 3-Isobutyl-1-methyl-1H,3H-purin-2,6-dion, 2,6-Dihydroxy-1-methyl-3-isobutylpurin

13347-100 13347-250 13347-500
CAYM13347-100EA 50 EUR
CAYM13347-100 CAYM13347-250 CAYM13347-500
3-Isobutyl-1-methylxanthin
3-Isobutyl-1-methylxanthin
Formel: C₁₀H₁₄N₄O₂
Schmelzpunkt: 200…201 °C
MDL: MFCD00005584
CAS-Nummer: 28822-58-4
EINECS: 249-259-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR