Lansoprazol

Lieferant: Cayman Chemical

Synonyme: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

13627-500 13627-1 13627-5 13627-10
CAYM13627-500EA 52 EUR
CAYM13627-500 CAYM13627-1 CAYM13627-5 CAYM13627-10
Lansoprazol
Lansoprazol
Formel: C₁₆H₁₄F₃N₃O₂S
Molecular Weight: 369.36 g/mol
MDL: MFCD00866873
CAS-Nummer: 103577-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR