(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on

Lieferant: Cayman Chemical

14173-1 14173-5 14173-10
CAYM14173-1EA 50 EUR
CAYM14173-1 CAYM14173-5 CAYM14173-10
(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on
(±)-Naringenin
Formel: C₁₅H₁₂O₅
MDL: MFCD00006844
CAS-Nummer: 480-41-1
EINECS: 207-550-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR