Rivastigmine-L-tartrate ≥95%, kristalliner Feststoff
Lieferant: Cayman Chemical
|
Gefahr
|
Synonyme:
Rivastigmine tartrate, (S)-3-(1-(Dimethylamino)ethyl)phenyl ethyl(methyl)carbamate (2R,3R)-2,3-dihydroxysuccinate, N-Ethyl-N-methylcarbamic Acid 3-[(S)-1-(Dimethylamino)ethyl]phenyl Ester L-Tartrate, 3-[(S)-1-(Dimethylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate L-Tartrate
Rivastigmine is an irreversible inhibitor of acetylcholinesterase (IC50=4,15 µm) and butyrylcholinesterase (IC50=37 nM).
It acts by covalently modifying a serine residue in the active site by carbamoylation. Rivastigmine has found use in modifying the course of neurodegenerative diseases, including Parkinson’s disease and Alzheimer’s disease, as it stabilises or reduces the rate of decline in certain cognitive functions.
Formel:
C₁₄H₂₂N₂O₂·C₄H₆O₆ Lagertemperatur: Gefrierschrank |
MDL:
MFCD03700731 CAS-Nummer: 129101-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...