TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)

Lieferant: Cayman Chemical
Gefahr

Synonyme: TCEP HCl

14329-1
CAYM14329-1EA 59 EUR
CAYM14329-1
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
TCEP-HCl (Tris(2-carboxyethyl)phosphin Hydrochlorid)
Formel: C₉H₁₆ClO₆P
Molecular Weight: 286,65 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00145469
CAS-Nummer: 51805-45-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR