Acridinorange

Lieferant: Cayman Chemical

Synonyme: Brilliant Acridine Orange E, 3,6-Bis(dimethylamino)acridine, Acridine Orange base, Solvent Orange 15, 3,6-Bis(dimethylamino)acridine N,N,N',N'-Tetramethyl-3,6-acridinediamine, Acridine orange free base, Acridine Orange Stain

14338-5 14338-10 14338-25
CAYM14338-5EA 83 EUR
CAYM14338-5 CAYM14338-10 CAYM14338-25
Acridinorange
Acridinorange
Formel: C₁₇H₁₉N₃
MDL: MFCD00005029
CAS-Nummer: 494-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR