Phloretin

Lieferant: Cayman Chemical

14452-250 14452-500 14452-1
CAYM14452-250EA 55 EUR
CAYM14452-250 CAYM14452-500 CAYM14452-1
Phloretin
Phloretin
Formel: C₁₅H₁₄O₅
MDL: MFCD00002288
CAS-Nummer: 60-82-2
EINECS: 200-488-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR